Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01426821_g1
          See other TRMT1 GE Assays ›
          Gene Symbol:
          Trmt1
          Gene Name:
          tRNA methyltransferase 1 homolog (S. cerevisiae)
          Gene Aliases:
          RGD1305992
          Chromosome Location:
          Chr.19: 25798263 - 25813458 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01426821_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Trmt1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001013870.1 NP_001013892.1 15 - 16 2291 138
          XM_006255236.2 XP_006255298.1 17 - 18 4079 138
          XM_008772365.1 XP_008770587.1 17 - 18 4155 138
          XM_008772366.1 XP_008770588.1 16 - 17 3940 138
          GenBank mRNA BC085126.1 - 15 - 16 2291 7132178 138

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1900000336.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Trmt1

          Panther Classification:

          Molecular Function -

          RNA methyltransferase

          Gene Ontology Categories:

          Function(s) Process(es)

          tRNA methylation
          tRNA (guanine-N2-)-methyltransferase activity
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P503964 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 273266 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 276757 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 239170 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 394196 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 88371 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 91583 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 430752 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 130643 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P503964 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P503964 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P503964 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P503964 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 62814 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10810083 - -
          Affymetrix Rat Gene 1.0 ST Array 10810079 - -
          Affymetrix Rat Gene 1.0 ST Array 10810082 - -
          Affymetrix Rat Gene 1.0 ST Array 10810084 - -
          Affymetrix Rat Gene 1.0 ST Array 10810073 - -
          Affymetrix Rat Gene 1.0 ST Array 10810077 - -
          Affymetrix Rat Gene 1.0 ST Array 10810081 - -
          Affymetrix Rat Gene 1.0 ST Array 10810088 - -
          Affymetrix Rat Gene 1.0 ST Array 10810085 - -
          Affymetrix Rat Gene 1.0 ST Array 10810076 - -
          Affymetrix Rat Gene 1.0 ST Array 10810070 - -
          Affymetrix Rat Gene 1.0 ST Array 10810080 - -
          Affymetrix Rat Gene 1.0 ST Array 10810071 - -
          Affymetrix Rat Gene 1.0 ST Array 10810072 - -
          Affymetrix Rat Gene 1.0 ST Array 10810086 - -
          Affymetrix Rat Gene 1.0 ST Array 10810078 - -
          Affymetrix Rat Gene 1.0 ST Array 10810075 - -
          Affymetrix Rat Gene 1.0 ST Array 10810074 - -
          Affymetrix Rat Gene 1.1 ST Array 10810069 - -
          Rat Genome 230 2.0 Array 1389056_at - -
          Rat Genome 230 2.0 Array 1389056_PM_at - -
          Affymetrix HT Rat Focus Array 1389056_PM_at - -
          Affymetrix HT Rat Focus Array 1389056_at - -
          Affymetrix Rat Clariom S Assay PSR1900002621.rn.1 TC1900000336.rn.1 RTA TC
          Rat Expression Set 230 1389056_at - -
          Rat Expression Set 230 1389056_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10810069 - -
          Rat Genome U34 Set rc_AI012114_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21710219 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1361236 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline