Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cenpt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024257.1 | NP_001019428.1 | 5 - 6 | 535 | 69 | |
XM_006255494.2 | XP_006255556.1 | 6 - 7 | 653 | 69 | ||
XM_006255495.2 | XP_006255557.1 | 6 - 7 | 654 | 69 | ||
GenBank mRNA | BC093396.1 | - | 5 - 6 | 535 | 7388929 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |