Search
Search
| Gene Symbol: Dus2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106181.1 | NP_001099651.1 | 10 - 11 | 773 | 61 | |
| XM_006255479.1 | XP_006255541.1 | 6 - 7 | 931 | 61 | ||
| GenBank mRNA | BC158728.1 | - | 10 - 11 | 675 | 9091646 | 61 |
Set Membership: |
|