Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01427056_m1
          See other RAMP1 GE Assays ›
          Gene Symbol:
          Ramp1
          Gene Name:
          receptor (G protein-coupled) activity modifying protein 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.9: 98313632 - 98364206 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01427056_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ramp1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031645.1 NP_113833.1 2 - 3 227 72
          XM_006245473.2 XP_006245535.1 2 - 3 1091 72
          XM_006245474.2 XP_006245536.1 2 - 3 303 72
          GenBank mRNA AB030942.1 - 2 - 3 203 72
          AB042887.1 - 2 - 3 227 72
          AF181550.1 - 2 - 3 204 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0900001131.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ramp1

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          calcium ion transport
          intracellular protein transport
          signal transduction
          G-protein coupled receptor signaling pathway
          regulation of G-protein coupled receptor protein signaling pathway
          protein transport
          positive regulation of cAMP biosynthetic process
          receptor internalization
          positive regulation of protein glycosylation
          protein localization to plasma membrane
          G-protein coupled receptor binding
          receptor activity
          calcitonin receptor activity
          protein transporter activity
          coreceptor activity
          calcitonin receptor binding
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P159404 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P022090 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 923614 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 228150 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 548227 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 54370 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 526685 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 483262 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 863460 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 571383 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 84077 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 450129 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 385006 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 382879 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 786552 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 871488 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 41872 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 625307 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 1024 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 104135 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 644448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 811569 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 639207 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 240245 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 731155 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 874270 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 178111 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 82602 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 123109 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 24988 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 913640 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 280003 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 461072 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 106522 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 47040 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 20278 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 178970 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 133500 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 126829 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 95545 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P022090 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P159404 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P159404 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P022090 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P022090 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P159404 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1014241 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700502466 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 51657 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 63933 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 70363 - -
          Agilent Rat cDNA Microarray (G4105A) 700502466 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10925368 - -
          Affymetrix Rat Gene 1.0 ST Array 10925366 - -
          Affymetrix Rat Gene 1.0 ST Array 10925367 - -
          Affymetrix Rat Gene 1.0 ST Array 10925369 - -
          Affymetrix Rat Gene 1.0 ST Array 10925370 - -
          Affymetrix Rat Gene 1.1 ST Array 10925365 - -
          Rat Genome 230 2.0 Array 1367791_PM_at - -
          Rat Genome 230 2.0 Array 1367791_at - -
          Affymetrix HT Rat Focus Array 1367791_PM_at - -
          Affymetrix HT Rat Focus Array 1367791_at - -
          Affymetrix Rat Clariom S Assay PSR0900009785.rn.1 TC0900001131.rn.1 RTA TC
          Rat Expression Set 230 1367791_PM_at - -
          Rat Expression Set 230 1367791_at - -
          Affymetrix Rat Gene 1.0 ST Array 10925365 - -
          Rat Genome U34 Set rc_AI012429_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21484562 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1374522 - -
          RatRef-12 Expression BeadChip ILMN_49659 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline