Search
Search
| Gene Symbol: Wash | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127390.1 | NP_001120862.1 | 1306 | 115 | ||
| GenBank mRNA | BC088270.1 | - | 2 - 3 | 348 | 115 | |
| BC101888.1 | - | 5 - 6 | 662 | 115 | ||
| BC166761.1 | - | 1306 | 7930951 | 115 | ||
Set Membership: |
|