Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01427657_m1
          See other THAP4 GE Assays ›
          Gene Symbol:
          Thap4
          Gene Name:
          THAP domain containing 4
          Gene Aliases:
          -
          Chromosome Location:
          Chr.9: 100848597 - 100888107 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01427657_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Thap4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001005564.1 NP_001005564.1 5 - 6 1626 92
          XM_008767380.1 XP_008765602.1 4 - 5 1520 92
          GenBank mRNA BC081882.1 - 5 - 6 1626 7108379 92
          FQ217334.1 - 1 - 2 73 92

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0900002508.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Thap4

          Gene Ontology Categories:

          Process(es)

          DNA binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P199514 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 866502 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 336130 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 300200 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 182698 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 838394 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 519071 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 366761 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 718979 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 277446 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 389159 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 889826 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 272148 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 307785 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 938403 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 687198 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 206618 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 552322 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 201181 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 820598 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 677220 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 484637 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 64338 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 147409 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 92890 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 282429 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 775430 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 888715 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 22247 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 146805 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 137515 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 92459 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 17109 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 117328 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 165413 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P199514 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P199514 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P199514 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P199514 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 15783 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 36015 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 100291 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 50905 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10930071 - -
          Affymetrix Rat Gene 1.0 ST Array 10925645 - -
          Affymetrix Rat Gene 1.0 ST Array 10930074 - -
          Affymetrix Rat Gene 1.0 ST Array 10930073 - -
          Affymetrix Rat Gene 1.0 ST Array 10930072 - -
          Affymetrix Rat Gene 1.0 ST Array 10930075 - -
          Affymetrix Rat Gene 1.1 ST Array 10930070 - -
          Affymetrix Rat Gene 1.1 ST Array 10925644 - -
          Rat Genome 230 2.0 Array 1371557_PM_at - -
          Rat Genome 230 2.0 Array 1371557_at - -
          Affymetrix HT Rat Focus Array 1371557_PM_at - -
          Affymetrix HT Rat Focus Array 1371557_at - -
          Affymetrix Rat Clariom S Assay PSR0900020661.rn.1 TC0900002508.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900020662.rn.1 TC0900002508.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900020663.rn.1 TC0900002508.rn.1 RTA TC
          Rat Expression Set 230 1371557_PM_at - -
          Rat Expression Set 230 1371557_at - -
          Affymetrix Rat Gene 1.0 ST Array 10930070 - -
          Rat Genome U34 Set rc_AA924144_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21745513 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1354690 - -
          RatRef-12 Expression BeadChip ILMN_66519 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline