Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01427713_m1
          See other MAN2A1 GE Assays ›
          Gene Symbol:
          Man2a1
          Gene Name:
          mannosidase, alpha, class 2A, member 1
          Gene Aliases:
          Mana2, Manno
          Chromosome Location:
          Chr.9: 112293388 - 112451677 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01427713_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Man2a1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012979.2 NP_037111.2 13 - 14 3091 62
          GenBank mRNA AY325162.1 - 1268 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0900001261.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          in utero embryonic development
          liver development
          mannose metabolic process
          protein glycosylation
          N-glycan processing
          mitochondrion organization
          vacuole organization
          respiratory gaseous exchange
          lung alveolus development
          positive regulation of neurogenesis
          retina morphogenesis in camera-type eye
          mannosyl-oligosaccharide 1,3-1,6-alpha-mannosidase activity
          zinc ion binding
          mannosidase activity
          hydrolase activity, hydrolyzing N-glycosyl compounds
          carbohydrate binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P707838 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P010174 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P135340 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 424874 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 636671 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 971954 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 934401 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 693810 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 545689 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 103821 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 525511 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 846137 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 808603 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 778241 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 258723 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 408749 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 405705 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 717258 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 691111 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 93914 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 390007 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 269351 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 527449 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 665858 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 767673 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 367088 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 932250 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 111891 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 54998 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 280603 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 606482 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 434756 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 563985 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 337263 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 785737 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 525771 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 376434 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 755635 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 973350 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 191248 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 786266 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 921341 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 963824 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 674019 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 48251 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 7788 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 205606 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 372100 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 116844 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 861760 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 831222 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 3040 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 791972 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 223116 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 285373 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 470966 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 888449 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 77322 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 528099 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 959174 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 122948 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 713176 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 575306 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 693306 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 341645 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 896454 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 399536 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 884754 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 576018 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 318990 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 375122 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 477033 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 313683 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 955398 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 232292 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 476239 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 253253 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 501426 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 792418 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 532057 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 229561 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 875356 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 127297 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 822447 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 897297 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 212965 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 506902 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 769605 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 327334 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 724344 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 468899 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 257807 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 796551 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 264718 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 284422 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 525507 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 538852 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 744957 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 694242 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 311768 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 682413 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 84387 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 377620 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 222916 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 787339 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 89044 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 914135 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 761295 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 959231 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 709128 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 174533 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 112167 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 53533 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 100522 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 172231 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 134910 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 33466 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 26491 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 70933 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 46573 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 25594 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 141993 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 174994 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 180282 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 87683 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 120481 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 9944 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 163904 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 175919 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P010174 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P135340 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P707838 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P010174 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P707838 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P135340 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P010174 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P135340 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P707838 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P320834 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P22715 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P16753 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P522209 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1038694 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 73611 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 78968 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 83975 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 60328 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 83279 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 56419 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10925803 - -
          Affymetrix Rat Gene 1.0 ST Array 10925786 - -
          Affymetrix Rat Gene 1.0 ST Array 10925807 - -
          Affymetrix Rat Gene 1.0 ST Array 10925800 - -
          Affymetrix Rat Gene 1.0 ST Array 10925788 - -
          Affymetrix Rat Gene 1.0 ST Array 10925792 - -
          Affymetrix Rat Gene 1.0 ST Array 10925798 - -
          Affymetrix Rat Gene 1.0 ST Array 10925789 - -
          Affymetrix Rat Gene 1.0 ST Array 10925793 - -
          Affymetrix Rat Gene 1.0 ST Array 10925806 - -
          Affymetrix Rat Gene 1.0 ST Array 10925790 - -
          Affymetrix Rat Gene 1.0 ST Array 10925804 - -
          Affymetrix Rat Gene 1.0 ST Array 10925799 - -
          Affymetrix Rat Gene 1.0 ST Array 10925784 - -
          Affymetrix Rat Gene 1.0 ST Array 10925795 - -
          Affymetrix Rat Gene 1.0 ST Array 10925794 - -
          Affymetrix Rat Gene 1.0 ST Array 10925805 - -
          Affymetrix Rat Gene 1.0 ST Array 10925801 - -
          Affymetrix Rat Gene 1.0 ST Array 10925802 - -
          Affymetrix Rat Gene 1.0 ST Array 10925785 - -
          Affymetrix Rat Gene 1.0 ST Array 10925791 - -
          Affymetrix Rat Gene 1.0 ST Array 10925787 - -
          Affymetrix Rat Gene 1.1 ST Array 10925783 - -
          Affymetrix HT Rat Focus Array 1388667_at - -
          Affymetrix HT Rat Focus Array 1383074_at - -
          Affymetrix HT Rat Focus Array 1383074_PM_at - -
          Affymetrix HT Rat Focus Array 1388667_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0900010743.rn.1 TC0900001261.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900010742.rn.1 TC0900001261.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10925783 - -
          Rat Genome U34 Set M24353_at - -
          Rat Genome U34 Set M24353_g_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21262719 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364314 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline