Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01427785_m1
          See other UGT1A5 GE Assays ›
          Gene Symbol:
          Ugt1a5
          Gene Name:
          UDP glucuronosyltransferase 1 family, polypeptide A5
          Gene Aliases:
          UDPGT 1-5, Ugt1, Ugt1a6
          Chromosome Location:
          Chr.9: 95256628 - 95302822 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01427785_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ugt1a5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001039549.1 NP_001034638.1 1 - 2 887 68
          GenBank mRNA AF461734.1 - 1 - 2 896 68
          AY435132.1 - 1 - 2 852 68

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          glycosyltransferase

          Gene Ontology Categories:

          Function(s) Process(es)

          liver development
          flavonoid biosynthetic process
          response to organic cyclic compound
          cellular response to hormone stimulus
          flavonoid glucuronidation
          xenobiotic glucuronidation
          cellular response to ethanol
          cellular response to glucocorticoid stimulus
          glucuronosyltransferase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10925183 - -
          Affymetrix Rat Gene 1.0 ST Array 10925178 - -
          Affymetrix Rat Gene 1.0 ST Array 10925176 - -
          Affymetrix Rat Gene 1.0 ST Array 10925167 - -
          Affymetrix Rat Gene 1.0 ST Array 10925177 - -
          Affymetrix Rat Gene 1.0 ST Array 10925182 - -
          Affymetrix Rat Gene 1.0 ST Array 10925179 - -
          Affymetrix Rat Gene 1.0 ST Array 10925181 - -
          Affymetrix Rat Gene 1.0 ST Array 10925184 - -
          Affymetrix Rat Gene 1.0 ST Array 10925168 - -
          Affymetrix Rat Gene 1.0 ST Array 10925180 - -
          Affymetrix Rat Gene 1.1 ST Array 10925158 - -
          Rat Genome 230 2.0 Array 1387759_s_at - -
          Rat Genome 230 2.0 Array 1370613_s_at - -
          Rat Genome 230 2.0 Array 1387759_PM_s_at - -
          Rat Genome 230 2.0 Array 1370613_PM_s_at - -
          Rat Expression Set 230 1387759_PM_s_at - -
          Rat Expression Set 230 1387759_s_at - -
          Rat Expression Set 230 1370613_PM_s_at - -
          Rat Expression Set 230 1370613_s_at - -
          Affymetrix Rat Gene 1.0 ST Array 10925158 - -
          Rat Genome U34 Set J05132_s_at - -
          Rat Genome U34 Set J02612mRNA_s_at - -
          Rat Genome U34 Set D83796_s_at - -
          Rat Genome U34 Set S56937_s_at - -
          Rat Genome U34 Set D38069exon_s_at - -
          Rat Genome U34 Set J02612mRNA_s_at - -
          Rat Genome U34 Set J05132_s_at - -
          Rat Genome U34 Set D83796_s_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline