Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01428271_m1
          See other ZFP365 GE Assays ›
          Gene Symbol:
          Zfp365
          Gene Name:
          zinc finger protein 365
          Gene Aliases:
          Znf365
          Chromosome Location:
          Chr.20: 22060160 - 22083766 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01428271_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp365
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001025145.1 NP_001020316.1 4 - 5 1183 68
          GenBank mRNA BC087056.1 - 4 - 5 1183 7124000 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC2000000572.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zfp365

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P654145 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P073673 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 293035 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 53342 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 453277 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 356379 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 484673 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 760246 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 676162 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 861258 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 862139 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 7230 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 856064 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 701011 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 360717 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 441835 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 750594 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 130640 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 168930 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P073673 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P654145 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P073673 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P654145 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P073673 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P654145 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P654145 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 19535 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 61385 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 11612 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10829792 - -
          Affymetrix Rat Gene 1.0 ST Array 10829798 - -
          Affymetrix Rat Gene 1.0 ST Array 10829793 - -
          Affymetrix Rat Gene 1.0 ST Array 10829797 - -
          Affymetrix Rat Gene 1.0 ST Array 10829795 - -
          Affymetrix Rat Gene 1.0 ST Array 10829796 - -
          Affymetrix Rat Gene 1.0 ST Array 10829794 - -
          Affymetrix Rat Gene 1.1 ST Array 10829791 - -
          Rat Genome 230 2.0 Array 1393253_at - -
          Rat Genome 230 2.0 Array 1393253_PM_at - -
          Affymetrix HT Rat Focus Array 1393253_at - -
          Affymetrix HT Rat Focus Array 1393253_PM_at - -
          Affymetrix Rat Clariom S Assay PSR2000005180.rn.1 TC2000000572.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000005181.rn.1 TC2000000572.rn.1 RTA TC
          Rat Expression Set 230 1393253_PM_at - -
          Rat Expression Set 230 1393253_at - -
          Affymetrix Rat Gene 1.0 ST Array 10829791 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364903 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline