Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01428651_g1
          See other SLC39A7 GE Assays ›
          Gene Symbol:
          Slc39a7
          Gene Name:
          solute carrier family 39 (zinc transporter), member 7
          Gene Aliases:
          Ke4, RT1-Ke4, Rt1Ke4
          Chromosome Location:
          Chr.20: 3819416 - 3822788 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01428651_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc39a7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001008885.2 NP_001008885.1 6 - 7 1463 170
          NM_001164744.1 NP_001158216.1 7 - 8 1204 170
          GenBank mRNA BC079141.1 - 6 - 7 1463 7127902 170
          FQ212335.1 - 7 - 8 1171 170
          FQ212833.1 - 6 - 7 1490 170

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC2000001818.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc39a7

          Panther Classification:

          Molecular Function -

          primary active transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          metal ion transport
          transmembrane transport
          metal ion transmembrane transporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P286215 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 48052 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 811463 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 774315 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P286215 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P286215 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P286215 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P286215 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 45688 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10828392 - -
          Affymetrix Rat Gene 1.0 ST Array 10828391 - -
          Affymetrix Rat Gene 1.0 ST Array 10828386 - -
          Affymetrix Rat Gene 1.0 ST Array 10828388 - -
          Affymetrix Rat Gene 1.0 ST Array 10828390 - -
          Affymetrix Rat Gene 1.0 ST Array 10828387 - -
          Affymetrix Rat Gene 1.0 ST Array 10828393 - -
          Affymetrix Rat Gene 1.0 ST Array 10828389 - -
          Affymetrix Rat Gene 1.0 ST Array 10828385 - -
          Affymetrix Rat Gene 1.1 ST Array 10828384 - -
          Rat Genome 230 2.0 Array 1389089_PM_at - -
          Rat Genome 230 2.0 Array 1389089_at - -
          Affymetrix HT Rat Focus Array 1389089_at - -
          Affymetrix HT Rat Focus Array 1389089_PM_at - -
          Affymetrix Rat Clariom S Assay PSR2000014910.rn.1 TC2000001818.rn.1 RTA TC
          Rat Expression Set 230 1389089_PM_at - -
          Rat Expression Set 230 1389089_at - -
          Affymetrix Rat Gene 1.0 ST Array 10828384 - -
          Rat Genome U34 Set rc_AI013750_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21187134 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376697 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline