Search
Search
| Gene Symbol: Jmjd1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191719.1 | NP_001178648.1 | 12 - 13 | 5450 | 70 | |
| XM_006256359.2 | XP_006256421.1 | 12 - 13 | 5504 | 70 | ||
| XM_006256360.2 | XP_006256422.1 | 11 - 12 | 5300 | 70 | ||
| XM_006256361.2 | XP_006256423.1 | 12 - 13 | 5527 | 70 | ||
| XM_006256362.2 | XP_006256424.1 | 12 - 13 | 5419 | 70 | ||
| XM_006256363.1 | XP_006256425.1 | 8 - 9 | 4861 | 70 | ||
| XM_006256364.1 | XP_006256426.1 | 12 - 13 | 5413 | 70 | ||
| XM_006256365.1 | XP_006256427.1 | 7 - 8 | 4719 | 70 | ||
Set Membership: |
|