Search
Search

Col11a2
LOC100911886| Gene Symbol: Col11a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212528.1 | NP_997693.1 | 16 - 17 | 1674 | 139 | |
| XM_006255928.2 | XP_006255990.1 | 18 - 19 | 2780 | 139 | ||
| XM_006255929.2 | XP_006255991.1 | 15 - 16 | 2459 | 139 | ||
| XM_008772720.1 | XP_008770942.1 | 20 - 21 | 2904 | 139 | ||
| Gene Symbol: LOC100911886 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008772788.1 | XP_008771010.1 | 20 - 21 | 1783 | 139 | |
Set Membership: |
|