Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01429616_m1
          See other HERC4 GE Assays ›
          Gene Symbol:
          Herc4
          Gene Name:
          HECT and RLD domain containing E3 ubiquitin protein ligase 4
          Gene Aliases:
          -
          Chromosome Location:
          Chr.20: 26755935 - 26831267 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01429616_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Herc4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001012074.1 NP_001012074.1 23 - 24 3105 129
          XM_006256134.2 XP_006256196.1 22 - 23 3087 129
          XM_006256135.2 XP_006256197.1 22 - 23 3094 129
          XM_006256136.2 XP_006256198.1 21 - 22 2938 129
          GenBank mRNA BC087104.1 - 23 - 24 3105 7192894 129
          FQ218928.1 - 5 - 6 538 129

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC2000000610.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Herc4

          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          spermatogenesis
          cell differentiation
          protein ubiquitination involved in ubiquitin-dependent protein catabolic process
          ubiquitin-protein transferase activity
          ligase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P458075 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 531063 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 294892 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 6053 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 434214 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 116141 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 772507 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 972678 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 250349 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 643623 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 618407 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 184647 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 385782 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 671682 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721542 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 360828 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 278791 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 585368 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 311149 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 92251 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 30849 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 217553 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 369372 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 452820 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 245512 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 963993 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 339434 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 204286 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 836951 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 201847 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 404123 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 354020 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 537746 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 956762 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 962995 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 472924 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 335869 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 277246 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 501680 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 439733 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 916488 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 130833 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 185441 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 839162 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 606408 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 526099 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 286355 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 771000 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 26100 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 744418 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 698650 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 269715 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 291318 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 138685 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 86246 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 102304 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 12258 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 134047 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 10325 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 124786 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 164086 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P458075 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P458075 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P458075 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P19167 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 29131 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 76938 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 60072 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 64887 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 77872 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10829905 - -
          Affymetrix Rat Gene 1.0 ST Array 10829910 - -
          Affymetrix Rat Gene 1.0 ST Array 10829900 - -
          Affymetrix Rat Gene 1.0 ST Array 10832904 - -
          Affymetrix Rat Gene 1.0 ST Array 10829924 - -
          Affymetrix Rat Gene 1.0 ST Array 10829908 - -
          Affymetrix Rat Gene 1.0 ST Array 10829899 - -
          Affymetrix Rat Gene 1.0 ST Array 10829919 - -
          Affymetrix Rat Gene 1.0 ST Array 10829921 - -
          Affymetrix Rat Gene 1.0 ST Array 10829906 - -
          Affymetrix Rat Gene 1.0 ST Array 10829918 - -
          Affymetrix Rat Gene 1.0 ST Array 10829913 - -
          Affymetrix Rat Gene 1.0 ST Array 10829904 - -
          Affymetrix Rat Gene 1.0 ST Array 10829901 - -
          Affymetrix Rat Gene 1.0 ST Array 10829911 - -
          Affymetrix Rat Gene 1.0 ST Array 10829907 - -
          Affymetrix Rat Gene 1.0 ST Array 10829914 - -
          Affymetrix Rat Gene 1.0 ST Array 10829902 - -
          Affymetrix Rat Gene 1.0 ST Array 10829912 - -
          Affymetrix Rat Gene 1.0 ST Array 10829916 - -
          Affymetrix Rat Gene 1.0 ST Array 10829920 - -
          Affymetrix Rat Gene 1.0 ST Array 10829915 - -
          Affymetrix Rat Gene 1.0 ST Array 10829903 - -
          Affymetrix Rat Gene 1.0 ST Array 10829922 - -
          Affymetrix Rat Gene 1.0 ST Array 10829923 - -
          Affymetrix Rat Gene 1.0 ST Array 10829917 - -
          Affymetrix Rat Gene 1.0 ST Array 10829925 - -
          Affymetrix Rat Gene 1.0 ST Array 10829909 - -
          Affymetrix Rat Gene 1.1 ST Array 10832902 - -
          Affymetrix Rat Gene 1.1 ST Array 10829898 - -
          Rat Genome 230 2.0 Array 1373381_at - -
          Rat Genome 230 2.0 Array 1395012_PM_at - -
          Rat Genome 230 2.0 Array 1395012_at - -
          Rat Genome 230 2.0 Array 1373381_PM_at - -
          Affymetrix HT Rat Focus Array 1373381_PM_at - -
          Affymetrix HT Rat Focus Array 1395012_at - -
          Affymetrix HT Rat Focus Array 1373381_at - -
          Affymetrix HT Rat Focus Array 1395012_PM_at - -
          Affymetrix Rat Clariom S Assay PSR2000005345.rn.1 TC2000000610.rn.1 RTA TC
          Rat Expression Set 230 1373381_at - -
          Rat Expression Set 230 1373381_PM_at - -
          Rat Expression Set 230 1395012_PM_at - -
          Rat Expression Set 230 1395012_at - -
          Affymetrix Rat Gene 1.0 ST Array 10829898 - -
          Rat Genome U34 Set rc_AI029093_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21093028 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1375947 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376365 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline