Search
Search
| Gene Symbol: Ccnk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109672.1 | NP_001103142.1 | 5 - 6 | 524 | 59 | |
| XM_006240549.2 | XP_006240611.1 | 6 - 7 | 767 | 59 | ||
| XM_006240551.2 | XP_006240613.1 | 4 - 5 | 769 | 59 | ||
| GenBank mRNA | BC129121.1 | - | 5 - 6 | 524 | 7125896 | 59 |
Set Membership: |
|