Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ccnk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109672.1 | NP_001103142.1 | 7 - 8 | 687 | 68 | |
XM_006240549.2 | XP_006240611.1 | 8 - 9 | 930 | 68 | ||
XM_006240551.2 | XP_006240613.1 | 6 - 7 | 932 | 68 | ||
GenBank mRNA | BC129121.1 | - | 7 - 8 | 687 | 7125896 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |