Search
Search

Slc25a47| Gene Symbol: Slc25a47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001509.1 | NP_001001509.1 | 4 - 5 | 368 | 98 | |
| GenBank mRNA | AY603424.1 | - | 4 - 5 | 368 | 98 | |
| BC089874.1 | - | 4 - 5 | 372 | 7325453 | 98 | |
| FQ210765.1 | - | 4 - 5 | 428 | 98 | ||
Set Membership: |
|