Search
Search

Mthfd1l| Gene Symbol: Mthfd1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108462.2 | NP_001101932.2 | 2 - 3 | 414 | 60 | |
| GenBank mRNA | BC166462.1 | - | 2 - 3 | 414 | 9030223 | 60 |
Set Membership: |
|