Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01430599_m1
          See other CHST7 GE Assays ›
          Gene Symbol:
          Chst7
          Gene Name:
          carbohydrate (N-acetylglucosamine 6-O) sulfotransferase 7
          Gene Aliases:
          Gn6st-4
          Chromosome Location:
          Chr.X: 2623018 - 2657155 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01430599_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Chst7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_207600.2 NP_997483.1 1 - 2 1605 93
          GenBank mRNA BC111079.1 - 1 - 2 1605 7387313 93

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0X00001392.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Chst7

          Panther Classification:

          Molecular Function -

          transferase

          Gene Ontology Categories:

          Function(s) Process(es)

          carbohydrate metabolic process
          N-acetylglucosamine metabolic process
          sulfur compound metabolic process
          chondroitin sulfate biosynthetic process
          N-acetylglucosamine 6-O-sulfotransferase activity
          chondroitin 6-sulfotransferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P062608 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 701518 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 11349 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 907967 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 722202 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 473987 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 602993 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 409322 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 706622 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 956842 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 800893 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 477369 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 305499 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 733058 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 855825 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 317795 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 837466 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 696419 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 884924 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 165710 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 290213 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 87229 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 28787 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 178982 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 88906 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P062608 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P062608 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P062608 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P520005 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 11388 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 25017 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 12477 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10936658 - -
          Affymetrix Rat Gene 1.0 ST Array 10936657 - -
          Affymetrix Rat Gene 1.0 ST Array 10936656 - -
          Affymetrix Rat Gene 1.0 ST Array 10936659 - -
          Affymetrix Rat Gene 1.1 ST Array 10936655 - -
          Affymetrix Rat Clariom S Assay PSR0X00008861.rn.1 TC0X00001392.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10936655 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21596807 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1360699 - -
          RatRef-12 Expression BeadChip ILMN_54320 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline