Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01430882_g1
          See other DDX3X GE Assays ›
          Gene Symbol:
          Ddx3x
          Gene Name:
          DEAD (Asp-Glu-Ala-Asp) box helicase 3, X-linked
          Gene Aliases:
          -
          Chromosome Location:
          Chr.X: 10400363 - 10414020 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01430882_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ddx3x
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001108246.1 NP_001101716.1 13 - 14 1592 61
          XM_006256652.2 XP_006256714.1 15 - 16 1888 61
          XM_006256653.2 XP_006256715.1 15 - 16 1885 61
          GenBank mRNA FQ211849.1 - 15 - 16 1867 61

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P014918 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P357579 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721114 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 5803 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 455490 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 79857 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 444177 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 95511 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 226273 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 768855 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 42755 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 115912 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P014918 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P357579 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P014918 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P357579 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P357579 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P014918 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P199274 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P357579 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10936771 - -
          Affymetrix Rat Gene 1.0 ST Array 10936765 - -
          Affymetrix Rat Gene 1.0 ST Array 10936767 - -
          Affymetrix Rat Gene 1.0 ST Array 10936762 - -
          Affymetrix Rat Gene 1.0 ST Array 10936758 - -
          Affymetrix Rat Gene 1.0 ST Array 10936763 - -
          Affymetrix Rat Gene 1.0 ST Array 10936770 - -
          Affymetrix Rat Gene 1.0 ST Array 10936772 - -
          Affymetrix Rat Gene 1.0 ST Array 10936757 - -
          Affymetrix Rat Gene 1.0 ST Array 10936760 - -
          Affymetrix Rat Gene 1.0 ST Array 10936764 - -
          Affymetrix Rat Gene 1.0 ST Array 10936769 - -
          Affymetrix Rat Gene 1.0 ST Array 10936766 - -
          Affymetrix Rat Gene 1.0 ST Array 10936754 - -
          Affymetrix Rat Gene 1.0 ST Array 10936768 - -
          Affymetrix Rat Gene 1.0 ST Array 10936761 - -
          Affymetrix Rat Gene 1.0 ST Array 10936759 - -
          Affymetrix Rat Gene 1.0 ST Array 10936756 - -
          Affymetrix Rat Gene 1.1 ST Array 10936755 - -
          Affymetrix Rat Gene 1.1 ST Array 10936753 - -
          Rat Genome 230 2.0 Array 1372242_PM_at - -
          Rat Genome 230 2.0 Array 1385214_at - -
          Rat Genome 230 2.0 Array 1386535_at - -
          Rat Genome 230 2.0 Array 1385214_PM_at - -
          Rat Genome 230 2.0 Array 1386535_PM_at - -
          Rat Genome 230 2.0 Array 1372242_at - -
          Affymetrix HT Rat Focus Array 1372242_at - -
          Affymetrix HT Rat Focus Array 1385214_at - -
          Affymetrix HT Rat Focus Array 1386535_PM_at - -
          Affymetrix HT Rat Focus Array 1385214_PM_at - -
          Affymetrix HT Rat Focus Array 1372242_PM_at - -
          Affymetrix HT Rat Focus Array 1386535_at - -
          Affymetrix Rat Clariom S Assay PSR0X00009026.rn.1 TC0X00001435.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00009027.rn.1 TC0X00001435.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00009025.rn.1 TC0X00001435.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00009024.rn.1 TC0X00001435.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00009028.rn.1 TC0X00001435.rn.1 RTA TC
          Rat Expression Set 230 1372242_PM_at - -
          Rat Expression Set 230 1372242_at - -
          Rat Expression Set 230 1385214_at - -
          Rat Expression Set 230 1386535_PM_at - -
          Rat Expression Set 230 1385214_PM_at - -
          Rat Expression Set 230 1386535_at - -
          Affymetrix Rat Gene 1.0 ST Array 10936755 - -
          Affymetrix Rat Gene 1.0 ST Array 10936753 - -
          Rat Genome U34 Set rc_AA892042_at - -
          Rat Genome U34 Set rc_AI230369_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21613818 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_2038837 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-8ltcg:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline