Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01431032_m1
          See other SLC10A7 GE Assays ›
          Gene Symbol:
          Slc10a7
          Gene Name:
          solute carrier family 10, member 7
          Gene Aliases:
          P7
          Chromosome Location:
          Chr.19: 32857984 - 33080772 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01431032_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc10a7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001010948.1 NP_001010948.1 9 - 10 777 65
          GenBank mRNA AY825929.1 - 9 - 10 777 65
          FQ211514.1 - 965 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1900001305.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Transporter Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc10a7

          Panther Classification:

          Molecular Function -

          secondary carrier transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          sodium ion transport
          transmembrane transport
          symporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P139463 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 967274 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 114542 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 882942 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 577602 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 905578 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 666375 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 101476 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 413847 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 279659 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 917987 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 232570 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 677615 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 888223 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 247607 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 910611 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 727701 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 291956 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 215794 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 358635 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 600887 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 872903 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 482215 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 373122 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 955112 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 584881 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 346869 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 658599 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 272241 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 588477 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 878573 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 464220 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 556046 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 559572 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 180321 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 758026 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 959171 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 805283 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 317584 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 904466 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 201466 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 952351 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 30678 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 620053 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 566246 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 213773 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 633573 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 852316 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 864869 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 230971 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 677751 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 618778 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 130613 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 925736 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 934423 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 854450 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 384558 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 505468 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 702829 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 823145 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 126102 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 50602 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 577830 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 562823 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 367835 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 499628 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 160855 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 826974 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 140376 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 597266 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 420644 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 712811 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 389609 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 725014 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 818948 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 451284 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 323354 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 565078 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 43707 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 147690 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 155856 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 175693 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 718096 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 613082 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 424084 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 303091 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 261820 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 573946 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 296661 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 313728 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 874936 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 555168 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 424773 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 735631 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 47701 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 17438 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 474625 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 485576 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 177789 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 335826 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 106493 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 433205 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 706119 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 92096 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 841868 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 622772 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 961746 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 286596 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 542835 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 606184 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 287978 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 282052 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 569670 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 851566 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 6812 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 108429 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 228081 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 95541 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 505630 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 552097 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 958808 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 41549 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 457176 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 369274 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 75632 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 167367 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 338871 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 373508 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 500767 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 345776 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 308799 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 788839 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 904179 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 697047 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 357446 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 511707 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 819472 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 146354 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 36897 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 71311 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 95116 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 166854 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 24386 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 70158 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 43814 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 96536 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 79886 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 88102 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 96487 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 168339 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 115657 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 80186 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 39039 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 51209 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 40299 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 131143 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 100163 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 21799 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 107228 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 87382 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P139463 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P139463 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P139463 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P471136 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 22069 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 10962 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 92443 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 99901 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 96282 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 66939 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 94296 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 48226 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 69613 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 95381 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 103374 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 12167 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 79701 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 69696 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10810506 - -
          Affymetrix Rat Gene 1.0 ST Array 10807009 - -
          Affymetrix Rat Gene 1.0 ST Array 10807012 - -
          Affymetrix Rat Gene 1.0 ST Array 10810505 - -
          Affymetrix Rat Gene 1.0 ST Array 10807011 - -
          Affymetrix Rat Gene 1.0 ST Array 10810508 - -
          Affymetrix Rat Gene 1.0 ST Array 10807008 - -
          Affymetrix Rat Gene 1.0 ST Array 10810504 - -
          Affymetrix Rat Gene 1.0 ST Array 10807010 - -
          Affymetrix Rat Gene 1.0 ST Array 10807013 - -
          Affymetrix Rat Gene 1.0 ST Array 10810507 - -
          Affymetrix Rat Gene 1.1 ST Array 10810503 - -
          Affymetrix Rat Gene 1.1 ST Array 10807007 - -
          Rat Genome 230 2.0 Array 1382758_at - -
          Rat Genome 230 2.0 Array 1382758_PM_at - -
          Affymetrix HT Rat Focus Array 1382758_at - -
          Affymetrix HT Rat Focus Array 1382758_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1900010498.rn.1 TC1900001305.rn.1 RTA TC
          Rat Expression Set 230 1382758_at - -
          Rat Expression Set 230 1382758_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10807007 - -
          Affymetrix Rat Gene 1.0 ST Array 10810503 - -
          Rat Genome U34 Set rc_AI179697_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20912972 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1650893 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline