Search
Search

RGD1564093| Gene Symbol: RGD1564093 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105946.1 | NP_001099416.1 | 1 - 2 | 406 | 129 | |
| GenBank mRNA | FQ231178.1 | - | 1 - 2 | 345 | 129 | |
| FQ235293.1 | - | 1 - 2 | 129 | |||
Set Membership: |
|