Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01431212_g1
          See other GCSH GE Assays ›
          Gene Symbol:
          Gcsh
          Gene Name:
          glycine cleavage system protein H (aminomethyl carrier)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.19: 49522054 - 49532811 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01431212_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gcsh
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_133598.2 NP_598282.2 1 - 2 226 74
          GenBank mRNA BC088114.1 - 1 - 2 163 7321648 74
          CB609750.1 - 1 - 2 252 74
          Y17321.1 - 1 - 2 230 74

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gcsh

          Gene Ontology Categories:

          Function(s) Process(es)

          glycine decarboxylation via glycine cleavage system
          methylation
          aminomethyltransferase activity
          enzyme binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P262401 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P555418 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 288504 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 949934 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 393793 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 799657 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 27812 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 322625 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 12968 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P555418 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P262401 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P262401 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P555418 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P262401 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P555418 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P13225 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P262401 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700514561 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 37298 - -
          Agilent Rat cDNA Microarray (G4105A) 700514561 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10811298 - -
          Affymetrix Rat Gene 1.0 ST Array 10811295 - -
          Affymetrix Rat Gene 1.0 ST Array 10811294 - -
          Affymetrix Rat Gene 1.0 ST Array 10811296 - -
          Affymetrix Rat Gene 1.0 ST Array 10811297 - -
          Affymetrix Rat Gene 1.1 ST Array 10811293 - -
          Rat Genome 230 2.0 Array 1367908_at - -
          Rat Genome 230 2.0 Array 1367908_PM_at - -
          Affymetrix HT Rat Focus Array 1367908_PM_at - -
          Affymetrix HT Rat Focus Array 1367908_at - -
          Affymetrix Rat Clariom S Assay PSR1900012538.rn.1 TC1900001544.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900012539.rn.1 TC1900001544.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900012537.rn.1 TC1900001544.rn.1 RTA TC
          Rat Expression Set 230 1367908_PM_at - -
          Rat Expression Set 230 1367908_at - -
          Affymetrix Rat Gene 1.0 ST Array 10811293 - -
          Rat Genome U34 Set rc_AI103550_at - -
          Rat Genome U34 Set rc_AI172253_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21302935 - -
          ABI Rat Genome Survey Microarray 21850199 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_2039266 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline