Search
Search
| Gene Symbol: Dpep3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008383.1 | NP_001008384.1 | 7 - 8 | 1124 | 69 | |
| XM_006255517.2 | XP_006255579.1 | 8 - 9 | 1145 | 69 | ||
| XM_008772521.1 | XP_008770743.1 | 8 - 9 | 1342 | 69 | ||
| GenBank mRNA | BC085826.1 | - | 7 - 8 | 1124 | 7131855 | 69 |
Set Membership: |
|