Search
Search
| Gene Symbol: Plcg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017168.1 | NP_058864.1 | 18 - 19 | 2132 | 54 | |
| XM_008772607.1 | XP_008770829.1 | 18 - 19 | 2148 | 54 | ||
| GenBank mRNA | J05155.1 | - | 18 - 19 | 2132 | 54 | |
Set Membership: |
|