Search
Search

Exoc3l1| Gene Symbol: Exoc3l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106178.1 | NP_001099648.1 | 9 - 10 | 1745 | 62 | |
| XM_006255456.2 | XP_006255518.1 | 10 - 11 | 2162 | 62 | ||
| XM_006255457.2 | XP_006255519.1 | 10 - 11 | 2498 | 62 | ||
| XM_006255460.2 | XP_006255522.1 | 9 - 10 | 1676 | 62 | ||
| XM_006255461.2 | XP_006255523.1 | 9 - 10 | 1625 | 62 | ||
| XM_006255463.1 | XP_006255525.1 | 7 - 8 | 1370 | 62 | ||
| XM_008772478.1 | XP_008770700.1 | 10 - 11 | 2381 | 62 | ||
Set Membership: |
|