Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01432107_g1
          See other ACD GE Assays ›
          Gene Symbol:
          Acd
          Gene Name:
          adrenocortical dysplasia homolog (mouse)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.19: 37665295 - 37668041 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01432107_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Acd
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001037193.1 NP_001032270.1 10 - 11 1223 63
          XM_008772496.1 XP_008770718.1 10 - 11 1295 63
          GenBank mRNA BC099222.1 - 10 - 11 1223 7388035 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1900001354.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Acd

          Gene Ontology Categories:

          Function(s) Process(es)

          telomere maintenance
          skeletal system development
          urogenital system development
          intracellular protein transport
          telomere capping
          embryonic limb morphogenesis
          protection from non-homologous end joining at telomere
          telomere assembly
          negative regulation of telomere maintenance via telomerase
          segmentation
          positive regulation of telomerase activity
          positive regulation of single-stranded telomeric DNA binding
          protein localization to chromosome, telomeric region
          DNA binding
          DNA polymerase binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P810179 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 908484 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 492648 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P810179 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P810179 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P810179 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P810179 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1048388 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 102608 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10810671 - -
          Affymetrix Rat Gene 1.0 ST Array 10810664 - -
          Affymetrix Rat Gene 1.0 ST Array 10810669 - -
          Affymetrix Rat Gene 1.0 ST Array 10810675 - -
          Affymetrix Rat Gene 1.0 ST Array 10810663 - -
          Affymetrix Rat Gene 1.0 ST Array 10810667 - -
          Affymetrix Rat Gene 1.0 ST Array 10810666 - -
          Affymetrix Rat Gene 1.0 ST Array 10810674 - -
          Affymetrix Rat Gene 1.0 ST Array 10810672 - -
          Affymetrix Rat Gene 1.0 ST Array 10810670 - -
          Affymetrix Rat Gene 1.0 ST Array 10810665 - -
          Affymetrix Rat Gene 1.0 ST Array 10810673 - -
          Affymetrix Rat Gene 1.1 ST Array 10810662 - -
          Rat Genome 230 2.0 Array 1372429_at - -
          Rat Genome 230 2.0 Array 1372429_PM_at - -
          Affymetrix HT Rat Focus Array 1372429_PM_at - -
          Affymetrix HT Rat Focus Array 1372429_at - -
          Affymetrix Rat Clariom S Assay PSR1900010911.rn.1 TC1900001354.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900010912.rn.1 TC1900001354.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900010909.rn.1 TC1900001354.rn.1 RTA TC
          Rat Expression Set 230 1372429_PM_at - -
          Rat Expression Set 230 1372429_at - -
          Affymetrix Rat Gene 1.0 ST Array 10810662 - -
          Rat Genome U34 Set rc_AA899853_s_at - -
          Rat Genome U34 Set rc_AI012380_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21953972 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1367485 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline