Search
Search
| Gene Symbol: Kars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006967.1 | NP_001006968.1 | 3 - 4 | 397 | 68 | |
| XM_006255630.1 | XP_006255692.1 | 2 - 3 | 269 | 68 | ||
| GenBank mRNA | BC083652.1 | - | 3 - 4 | 397 | 7135143 | 68 |
Set Membership: |
|