Search
Search
| Gene Symbol: Cirh1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009640.1 | NP_001009640.1 | 2 - 3 | 193 | 84 | |
| XM_006255562.1 | XP_006255624.1 | 2 - 3 | 239 | 84 | ||
| XM_006255563.2 | XP_006255625.1 | 2 - 3 | 402 | 84 | ||
| XM_008772488.1 | XP_008770710.1 | 2 - 3 | 192 | 84 | ||
| GenBank mRNA | BC088461.1 | - | 2 - 3 | 193 | 7127580 | 84 |
Set Membership: |
|