Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01432898_m1
          See other TMEM170A GE Assays ›
          Gene Symbol:
          Tmem170a
          Gene Name:
          transmembrane protein 170A
          Gene Aliases:
          RGD1559505
          Chromosome Location:
          Chr.19: 44088568 - 44101365 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01432898_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmem170a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001134608.1 NP_001128080.1 2 - 3 351 131
          XM_008772528.1 XP_008770750.1 2 - 3 351 131

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1900001481.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P055895 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 431951 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 415298 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 515918 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 243596 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 162668 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P055895 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P055895 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P055895 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10811204 - -
          Affymetrix Rat Gene 1.0 ST Array 10811205 - -
          Affymetrix Rat Gene 1.1 ST Array 10811203 - -
          Rat Genome 230 2.0 Array 1393824_at - -
          Rat Genome 230 2.0 Array 1393824_PM_at - -
          Rat Genome 230 2.0 Array 1394815_PM_at - -
          Rat Genome 230 2.0 Array 1394815_at - -
          Affymetrix Rat Clariom S Assay PSR1900012206.rn.1 TC1900001481.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900012201.rn.1 TC1900001481.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900012202.rn.1 TC1900001481.rn.1 RTA TC
          Rat Expression Set 230 1393824_PM_at - -
          Rat Expression Set 230 1393824_at - -
          Rat Expression Set 230 1394815_PM_at - -
          Rat Expression Set 230 1394815_at - -
          Affymetrix Rat Gene 1.0 ST Array 10811203 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22418073 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1350160 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0