Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01433205_m1
          See other LEPR GE Assays ›
          Gene Symbol:
          Lepr
          Gene Name:
          leptin receptor
          Gene Aliases:
          Fa
          Chromosome Location:
          Chr.5: 120503475 - 120682281 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01433205_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lepr
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012596.1 NP_036728.1 18 - 19 2743 94
          GenBank mRNA AF287268.1 - 17 - 18 2669 94
          D84550.1 - 17 - 18 2669 94
          D84551.1 - 17 - 18 2669 94
          D85558.1 - 17 - 18 2689 94
          U52966.1 - 18 - 19 2743 94
          U60151.1 - 17 - 18 2669 94

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500001077.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lepr

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          ovulation from ovarian follicle
          response to hypoxia
          generation of precursor metabolites and energy
          G-protein coupled receptor signaling pathway
          female pregnancy
          response to nutrient
          cholesterol metabolic process
          cytokine-mediated signaling pathway
          leptin-mediated signaling pathway
          response to nicotine
          eating behavior
          positive regulation of MAPK cascade
          response to estrogen
          response to leptin
          negative regulation of gluconeogenesis
          negative regulation of hydrolase activity
          cellular response to organic substance
          cytokine receptor activity
          protein-hormone receptor activity
          peptide hormone binding
          identical protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P15244 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P082049 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 212702 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 411681 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 503860 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 758502 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 798364 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 248870 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 535993 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 191876 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 454987 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 515647 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 323812 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 112174 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 868719 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 564523 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 941071 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 209938 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 888246 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 247102 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 865325 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 505476 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 834603 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 901521 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 350734 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 970319 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 417350 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 108582 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 30802 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 614042 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 866413 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 372709 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 531728 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 366588 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 318591 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 433262 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 332679 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 820651 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 459694 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 395162 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 331378 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 244705 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 660982 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 84814 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 384002 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 428782 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 795187 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 704181 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 863363 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 915023 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 286868 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 584353 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 960333 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 351691 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 888316 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 25918 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 645487 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 191696 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 681342 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 508335 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 863427 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 830523 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 924346 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 714043 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 825165 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 209418 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 260126 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 265587 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 600289 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 600120 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 272279 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 884206 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605700 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 167391 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 687391 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 415489 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 197368 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 930992 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 389183 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 768757 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 183126 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 722817 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 486250 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 755613 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 600266 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 948481 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 196594 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 611496 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 136973 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 122767 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 173939 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 24687 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 147458 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 81562 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 95575 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 75658 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 25627 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 132656 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 83771 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 34824 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 147998 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 129218 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 71051 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 56095 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 88363 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 62711 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 134425 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 102546 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P15244 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P082049 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P082049 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P15244 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P15244 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P082049 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P469863 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 12078 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 96173 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 33041 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 98395 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 32447 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 100247 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 75665 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 79592 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 71626 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 63279 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10870305 - -
          Affymetrix Rat Gene 1.0 ST Array 10870299 - -
          Affymetrix Rat Gene 1.0 ST Array 10870308 - -
          Affymetrix Rat Gene 1.0 ST Array 10870314 - -
          Affymetrix Rat Gene 1.0 ST Array 10870291 - -
          Affymetrix Rat Gene 1.0 ST Array 10870311 - -
          Affymetrix Rat Gene 1.0 ST Array 10870309 - -
          Affymetrix Rat Gene 1.0 ST Array 10870312 - -
          Affymetrix Rat Gene 1.0 ST Array 10870315 - -
          Affymetrix Rat Gene 1.0 ST Array 10870307 - -
          Affymetrix Rat Gene 1.0 ST Array 10870304 - -
          Affymetrix Rat Gene 1.0 ST Array 10870303 - -
          Affymetrix Rat Gene 1.0 ST Array 10870313 - -
          Affymetrix Rat Gene 1.0 ST Array 10870298 - -
          Affymetrix Rat Gene 1.0 ST Array 10870300 - -
          Affymetrix Rat Gene 1.0 ST Array 10870302 - -
          Affymetrix Rat Gene 1.0 ST Array 10870310 - -
          Affymetrix Rat Gene 1.0 ST Array 10870306 - -
          Affymetrix Rat Gene 1.1 ST Array 10870289 - -
          Rat Genome 230 2.0 Array 1369739_at - -
          Rat Genome 230 2.0 Array 1388232_a_at - -
          Rat Genome 230 2.0 Array 1369739_PM_at - -
          Rat Genome 230 2.0 Array 1371223_PM_a_at - -
          Rat Genome 230 2.0 Array 1371222_PM_at - -
          Rat Genome 230 2.0 Array 1370604_PM_at - -
          Rat Genome 230 2.0 Array 1388232_PM_a_at - -
          Rat Genome 230 2.0 Array 1388260_a_at - -
          Rat Genome 230 2.0 Array 1370605_PM_s_at - -
          Rat Genome 230 2.0 Array 1370604_at - -
          Rat Genome 230 2.0 Array 1370605_s_at - -
          Rat Genome 230 2.0 Array 1388260_PM_a_at - -
          Rat Genome 230 2.0 Array 1371222_at - -
          Rat Genome 230 2.0 Array 1371223_a_at - -
          Affymetrix HT Rat Focus Array 1370604_PM_at - -
          Affymetrix HT Rat Focus Array 1370605_s_at - -
          Affymetrix HT Rat Focus Array 1388260_PM_a_at - -
          Affymetrix HT Rat Focus Array 1371223_a_at - -
          Affymetrix HT Rat Focus Array 1388232_a_at - -
          Affymetrix HT Rat Focus Array 1388232_PM_a_at - -
          Affymetrix HT Rat Focus Array 1370604_at - -
          Affymetrix HT Rat Focus Array 1369739_at - -
          Affymetrix HT Rat Focus Array 1370605_PM_s_at - -
          Affymetrix HT Rat Focus Array 1371222_PM_at - -
          Affymetrix HT Rat Focus Array 1371222_at - -
          Affymetrix HT Rat Focus Array 1371223_PM_a_at - -
          Affymetrix HT Rat Focus Array 1369739_PM_at - -
          Affymetrix HT Rat Focus Array 1388260_a_at - -
          Affymetrix Rat Clariom S Assay PSR0500006945.rn.1 TC0500001077.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500006946.rn.1 TC0500001077.rn.1 RTA TC
          Rat Expression Set 230 1388232_a_at - -
          Rat Expression Set 230 1370604_at - -
          Rat Expression Set 230 1370604_PM_at - -
          Rat Expression Set 230 1371223_PM_a_at - -
          Rat Expression Set 230 1369739_PM_at - -
          Rat Expression Set 230 1388260_a_at - -
          Rat Expression Set 230 1370605_PM_s_at - -
          Rat Expression Set 230 1371222_at - -
          Rat Expression Set 230 1370605_s_at - -
          Rat Expression Set 230 1388232_PM_a_at - -
          Rat Expression Set 230 1371223_a_at - -
          Rat Expression Set 230 1371222_PM_at - -
          Rat Expression Set 230 1388260_PM_a_at - -
          Rat Expression Set 230 1369739_at - -
          Affymetrix Rat Gene 1.0 ST Array 10870289 - -
          Rat Genome U34 Set U67207_s_at - -
          Rat Genome U34 Set U52966_at - -
          Rat Genome U34 Set rc_AA998983_at - -
          Rat Genome U34 Set U52966_at - -
          Rat Genome U34 Set U67207_s_at - -
          Rat Genome U34 Set rc_AA998983_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22015758 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373257 - -
          RatRef-12 Expression BeadChip ILMN_51321 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline