Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Mtap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047867.1 | NP_001041332.1 | 7 - 8 | 691 | 76 | |
XM_006238382.2 | XP_006238444.1 | 5 - 6 | 602 | 76 | ||
GenBank mRNA | AY325240.1 | - | 7 - 8 | 691 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |