Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01433275_m1
          See other CAAP1 GE Assays ›
          Gene Symbol:
          Caap1
          Gene Name:
          caspase activity and apoptosis inhibitor 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 113479416 - 113532960 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01433275_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Caap1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001034154.1 NP_001029326.1 5 - 6 559 94
          XM_006238400.2 XP_006238462.1 5 - 6 773 94
          XM_008763894.1 XP_008762116.1 5 - 6 557 94
          GenBank mRNA BC105829.1 - 5 - 6 559 7382013 94

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Caap1

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P087504 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 406428 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 187659 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 578069 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 963946 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 203706 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 340157 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 873993 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 868200 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 971891 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 965451 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 674106 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 553100 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 338210 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 887382 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 164977 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 36368 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 169724 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 814609 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 640842 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 923181 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 320620 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 768387 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 864426 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 354887 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 78318 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 417637 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 955023 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 87232 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 550745 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 825782 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 108338 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 503744 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 912782 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 29658 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 241859 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 779328 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 905245 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 418122 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 4522 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 170358 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 91225 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 33063 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 98830 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 136613 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 173646 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P087504 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P087504 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P087504 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P458281 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 89970 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 74498 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 67992 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 27553 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10878041 - -
          Affymetrix Rat Gene 1.0 ST Array 10878040 - -
          Affymetrix Rat Gene 1.0 ST Array 10878043 - -
          Affymetrix Rat Gene 1.0 ST Array 10869924 - -
          Affymetrix Rat Gene 1.0 ST Array 10878044 - -
          Affymetrix Rat Gene 1.0 ST Array 10878042 - -
          Affymetrix Rat Gene 1.0 ST Array 10878046 - -
          Affymetrix Rat Gene 1.1 ST Array 10878039 - -
          Affymetrix Rat Gene 1.1 ST Array 10869923 - -
          Rat Genome 230 2.0 Array 1380035_at - -
          Rat Genome 230 2.0 Array 1380035_PM_at - -
          Rat Genome 230 2.0 Array 1392591_PM_at - -
          Rat Genome 230 2.0 Array 1392591_at - -
          Affymetrix HT Rat Focus Array 1380035_PM_at - -
          Affymetrix HT Rat Focus Array 1392591_at - -
          Affymetrix HT Rat Focus Array 1380035_at - -
          Affymetrix HT Rat Focus Array 1392591_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0500022916.rn.1 TC0500003150.rn.1 RTA TC
          Rat Expression Set 230 1392591_at - -
          Rat Expression Set 230 1392591_PM_at - -
          Rat Expression Set 230 1380035_PM_at - -
          Rat Expression Set 230 1380035_at - -
          Affymetrix Rat Gene 1.0 ST Array 10878039 - -
          Rat Genome U34 Set rc_AI008418_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20861414 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1349802 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline