Search
Search

RGD1563815| Gene Symbol: RGD1563815 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127556.1 | NP_001121028.1 | 1 - 2 | 181 | 102 | |
| XM_008772898.1 | XP_008771120.1 | 1 - 2 | 203 | 102 | ||
Set Membership: |
|