Search
Search
| Gene Symbol: Vps26a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007740.1 | NP_001007741.1 | 6 - 7 | 740 | 77 | |
| XM_006256460.2 | XP_006256522.1 | 6 - 7 | 863 | 77 | ||
| XM_008772892.1 | XP_008771114.1 | 6 - 7 | 868 | 77 | ||
| GenBank mRNA | BC079150.1 | - | 6 - 7 | 740 | 7128580 | 77 |
| FQ211877.1 | - | 6 - 7 | 753 | 77 | ||
Set Membership: |
|