Search
Search
| Gene Symbol: Pald1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034128.2 | NP_001029300.2 | 16 - 17 | 2122 | 60 | |
| XM_006256446.2 | XP_006256508.1 | 17 - 18 | 2897 | 60 | ||
| XM_006256447.2 | XP_006256509.1 | 17 - 18 | 2333 | 60 | ||
Set Membership: |
|