Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01433599_m1
          See other MICU1 GE Assays ›
          Gene Symbol:
          Micu1
          Gene Name:
          mitochondrial calcium uptake 1
          Gene Aliases:
          Cbara1
          Chromosome Location:
          Chr.20: 29284848 - 29433617 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01433599_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Micu1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_199412.1 NP_955444.1 11 - 12 1310 103
          XM_006256420.2 XP_006256482.1 11 - 12 1425 103
          XM_006256421.2 XP_006256483.1 12 - 13 1418 103
          XM_006256422.2 XP_006256484.1 12 - 13 1412 103
          GenBank mRNA BC062075.1 - 11 - 12 1310 5599430 103

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC2000000651.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Micu1

          Panther Classification:

          Molecular Function -

          calcium-binding protein calmodulin-related

          Gene Ontology Categories:

          Function(s) Process(es)

          mitochondrial calcium ion transport
          calcium ion transmembrane import into mitochondrion
          protein homooligomerization
          mitochondrial calcium ion homeostasis
          positive regulation of mitochondrial calcium ion concentration
          calcium ion import
          calcium ion binding
          identical protein binding
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P068062 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P068062 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P068062 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P068062 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P147509 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P241479 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 10877 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 83271 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 4772 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 247 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 13742 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 76158 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 83104 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 30205 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 82262 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 44917 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 104864 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 56896 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 6156 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 38353 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 77740 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 65597 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 18332 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 2010 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10940597 - -
          Affymetrix Rat Gene 1.1 ST Array 10940597 - -
          Rat Genome 230 2.0 Array 1376715_PM_at - -
          Rat Genome 230 2.0 Array 1376715_at - -
          Affymetrix Rat Clariom S Assay PSR2000005714.rn.1 TC2000000651.rn.1 RTA TC
          Rat Expression Set 230 1376715_PM_at - -
          Rat Expression Set 230 1376715_at - -
          Affymetrix Rat Gene 1.0 ST Array 10940597 - -
          Rat Genome U34 Set rc_AA901065_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22314894 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1366384 - -
          RatRef-12 Expression BeadChip ILMN_64507 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-9kpsn:80/100.66.79.221:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0