Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01433611_m1
          See other HSF2 GE Assays ›
          Gene Symbol:
          Hsf2
          Gene Name:
          heat shock transcription factor 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.20: 38935820 - 38963444 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01433611_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hsf2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031694.2 NP_113882.1 7 - 8 861 82
          XM_006256505.2 XP_006256567.1 7 - 8 781 82
          XM_006256506.2 XP_006256568.1 7 - 8 1016 82
          GenBank mRNA AF172640.1 - 7 - 8 713 82
          BC086554.1 - 7 - 8 861 7302950 82

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC2000000764.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hsf2

          Panther Classification:

          Molecular Function -

          winged helix/forkhead transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription, DNA-templated
          spermatogenesis
          positive regulation of transcription from RNA polymerase II promoter
          RNA polymerase II regulatory region sequence-specific DNA binding
          RNA polymerase II intronic transcription regulatory region sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          protein homodimerization activity
          sequence-specific DNA binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P317287 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1024125 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 404702 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 528575 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 380618 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 584339 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 69021 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 910248 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 443188 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 340237 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 586521 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 393674 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 351377 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 205030 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 290114 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 654237 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 152598 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 154780 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 62000 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1024125 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P317287 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P317287 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1024125 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1024125 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P317287 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P317287 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1024125 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701578834 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 1207 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 9988 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 29676 - -
          Agilent Rat cDNA Microarray (G4105A) 701578834 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10830205 - -
          Affymetrix Rat Gene 1.0 ST Array 10830208 - -
          Affymetrix Rat Gene 1.0 ST Array 10830212 - -
          Affymetrix Rat Gene 1.0 ST Array 10830213 - -
          Affymetrix Rat Gene 1.0 ST Array 10830202 - -
          Affymetrix Rat Gene 1.0 ST Array 10830209 - -
          Affymetrix Rat Gene 1.0 ST Array 10830206 - -
          Affymetrix Rat Gene 1.0 ST Array 10830215 - -
          Affymetrix Rat Gene 1.0 ST Array 10830204 - -
          Affymetrix Rat Gene 1.0 ST Array 10830207 - -
          Affymetrix Rat Gene 1.0 ST Array 10830203 - -
          Affymetrix Rat Gene 1.0 ST Array 10830210 - -
          Affymetrix Rat Gene 1.0 ST Array 10830214 - -
          Affymetrix Rat Gene 1.1 ST Array 10830201 - -
          Rat Genome 230 2.0 Array 1391423_PM_at - -
          Rat Genome 230 2.0 Array 1382598_at - -
          Rat Genome 230 2.0 Array 1382598_PM_at - -
          Rat Genome 230 2.0 Array 1391423_at - -
          Rat Genome 230 2.0 Array 1391876_PM_at - -
          Rat Genome 230 2.0 Array 1387430_PM_at - -
          Rat Genome 230 2.0 Array 1391876_at - -
          Rat Genome 230 2.0 Array 1387430_at - -
          Affymetrix HT Rat Focus Array 1387430_at - -
          Affymetrix HT Rat Focus Array 1391876_PM_at - -
          Affymetrix HT Rat Focus Array 1387430_PM_at - -
          Affymetrix HT Rat Focus Array 1391876_at - -
          Affymetrix HT Rat Focus Array 1382598_at - -
          Affymetrix HT Rat Focus Array 1382598_PM_at - -
          Affymetrix Rat Clariom S Assay PSR2000006402.rn.1 TC2000000764.rn.1 RTA TC
          Rat Expression Set 230 1387430_PM_at - -
          Rat Expression Set 230 1387430_at - -
          Rat Expression Set 230 1382598_PM_at - -
          Rat Expression Set 230 1391423_at - -
          Rat Expression Set 230 1382598_at - -
          Rat Expression Set 230 1391876_PM_at - -
          Rat Expression Set 230 1391876_at - -
          Rat Expression Set 230 1391423_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10830201 - -
          Rat Genome U34 Set rc_AA944812_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1365182 - -
          RatRef-12 Expression BeadChip ILMN_57178 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline