Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01433678_m1
          See other PCBD1 GE Assays ›
          Gene Symbol:
          Pcbd1
          Gene Name:
          pterin-4 alpha-carbinolamine dehydratase/dimerization cofactor of hepatocyte nuclear factor 1 alpha
          Gene Aliases:
          DCoH, Pcbd, Pcd, Phs, TCF1
          Chromosome Location:
          Chr.20: 30689690 - 30696465 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01433678_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pcbd1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001007601.2 NP_001007602.1 3 - 4 254 90
          XM_006256414.1 XP_006256476.1 4 - 5 512 90
          GenBank mRNA DV724713.1 - 1 - 2 63 90
          FM060466.1 - 3 - 4 254 90
          FM067129.1 - 3 - 4 231 90
          M83740.1 - 3 - 4 450 90

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC2000000672.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pcbd1

          Panther Classification:

          Molecular Function -

          dehydratase

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          L-phenylalanine metabolic process
          tetrahydrobiopterin biosynthetic process
          biological_process
          regulation of protein homodimerization activity
          positive regulation of transcription, DNA-templated
          protein homotetramerization
          protein heterooligomerization
          oxidation-reduction process
          molecular_function
          transcription coactivator activity
          phenylalanine 4-monooxygenase activity
          4-alpha-hydroxytetrahydrobiopterin dehydratase activity
          identical protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P039414 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P039409 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 628905 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 771752 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 501573 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 613859 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 27184 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P039414 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P039409 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P039409 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P039414 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P039414 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P039409 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P555107 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 36748 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10830005 - -
          Affymetrix Rat Gene 1.0 ST Array 10830006 - -
          Affymetrix Rat Gene 1.0 ST Array 10830004 - -
          Affymetrix Rat Gene 1.0 ST Array 10830007 - -
          Affymetrix Rat Gene 1.1 ST Array 10830003 - -
          Rat Genome 230 2.0 Array 1372485_at - -
          Rat Genome 230 2.0 Array 1372485_PM_at - -
          Affymetrix HT Rat Focus Array 1372485_at - -
          Affymetrix HT Rat Focus Array 1372485_PM_at - -
          Affymetrix Rat Clariom S Assay PSR2000005912.rn.1 TC2000000672.rn.1 RTA TC
          Rat Expression Set 230 1372485_at - -
          Rat Expression Set 230 1372485_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10830003 - -
          Rat Genome U34 Set M83740_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20873850 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358139 - -
          RatRef-12 Expression BeadChip ILMN_55727 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline