Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01433744_g1
          See other ASF1A GE Assays ›
          Gene Symbol:
          Asf1a
          Gene Name:
          anti-silencing function 1A histone chaperone
          Gene Aliases:
          -
          Chromosome Location:
          Chr.20: 34894419 - 34909265 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01433744_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Asf1a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106389.1 NP_001099859.1 1 - 2 182 121

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          chromatin/chromatin-binding, or -regulatory protein

          Gene Ontology Categories:

          Function(s) Process(es)

          osteoblast differentiation
          DNA repair
          nucleosome assembly
          DNA replication-dependent nucleosome assembly
          DNA replication-independent nucleosome assembly
          muscle cell differentiation
          chromatin binding
          histone binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P010613 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P010618 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 742130 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 121052 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 393200 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 488839 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 634466 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 176103 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 474709 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 946144 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 596117 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 552467 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 175790 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 103643 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P010613 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P010613 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P010613 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P497339 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P503999 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P207881 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10830179 - -
          Affymetrix Rat Gene 1.0 ST Array 10830182 - -
          Affymetrix Rat Gene 1.0 ST Array 10830180 - -
          Affymetrix Rat Gene 1.0 ST Array 10830181 - -
          Affymetrix Rat Gene 1.1 ST Array 10830177 - -
          Rat Genome 230 2.0 Array 1379192_at - -
          Rat Genome 230 2.0 Array 1392768_at - -
          Rat Genome 230 2.0 Array 1383057_at - -
          Rat Genome 230 2.0 Array 1392768_PM_at - -
          Rat Genome 230 2.0 Array 1385616_PM_a_at - -
          Rat Genome 230 2.0 Array 1379192_PM_at - -
          Rat Genome 230 2.0 Array 1383057_PM_at - -
          Rat Genome 230 2.0 Array 1385616_a_at - -
          Affymetrix HT Rat Focus Array 1385616_a_at - -
          Affymetrix HT Rat Focus Array 1379192_PM_at - -
          Affymetrix HT Rat Focus Array 1392768_at - -
          Affymetrix HT Rat Focus Array 1383057_PM_at - -
          Affymetrix HT Rat Focus Array 1379192_at - -
          Affymetrix HT Rat Focus Array 1392768_PM_at - -
          Affymetrix HT Rat Focus Array 1385616_PM_a_at - -
          Affymetrix HT Rat Focus Array 1383057_at - -
          Affymetrix Rat Clariom S Assay PSR2000006300.rn.1 TC2000000737.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000006299.rn.1 TC2000000737.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000006293.rn.1 TC2000000737.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000006298.rn.1 TC2000000737.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000006296.rn.1 TC2000000737.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000006301.rn.1 TC2000000737.rn.1 RTA TC
          Rat Expression Set 230 1383057_at - -
          Rat Expression Set 230 1383057_PM_at - -
          Rat Expression Set 230 1379192_PM_at - -
          Rat Expression Set 230 1385616_PM_a_at - -
          Rat Expression Set 230 1385616_a_at - -
          Rat Expression Set 230 1379192_at - -
          Rat Expression Set 230 1392768_PM_at - -
          Rat Expression Set 230 1392768_at - -
          Rat Genome U34 Set rc_AI013266_at - -
          Rat Genome U34 Set rc_AA849843_at - -
          Rat Genome U34 Set rc_AA900944_at - -
          Rat Genome U34 Set rc_AI137136_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20868919 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1351513 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline