Search
Search

Col13a1| Gene Symbol: Col13a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109172.1 | NP_001102642.1 | 19 - 20 | 1487 | 56 | |
| XM_006256470.2 | XP_006256532.2 | 18 - 19 | 1478 | 56 | ||
| XM_006256479.2 | XP_006256541.1 | 18 - 19 | 1486 | 56 | ||
| XM_006256481.2 | XP_006256543.1 | 15 - 16 | 1356 | 56 | ||
| XM_008772904.1 | XP_008771126.1 | 18 - 19 | 1487 | 56 | ||
| XM_008772905.1 | XP_008771127.1 | 17 - 18 | 1460 | 56 | ||
| XM_008772906.1 | XP_008771128.1 | 18 - 19 | 1487 | 56 | ||
| XM_008772907.1 | XP_008771129.1 | 18 - 19 | 1487 | 56 | ||
| XM_008772908.1 | XP_008771130.1 | 17 - 18 | 1451 | 56 | ||
| XM_008772909.1 | XP_008771131.1 | 16 - 17 | 1424 | 56 | ||
| XM_008772910.1 | XP_008771132.1 | 17 - 18 | 1421 | 56 | ||
| XM_008772911.1 | XP_008771133.1 | 18 - 19 | 1488 | 56 | ||
| XM_008772912.1 | XP_008771134.1 | 15 - 16 | 1373 | 56 | ||
| GenBank mRNA | BC162018.1 | - | 15 - 16 | 1121 | 9098304 | 56 |
Set Membership: |
|