Search
Search

MGC112715| Gene Symbol: MGC112715 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044300.1 | NP_001037765.1 | 1 - 2 | 175 | 120 | |
| XM_006256426.2 | XP_006256488.1 | 1 - 2 | 226 | 120 | ||
| GenBank mRNA | BC098723.1 | - | 1 - 2 | 175 | 7377326 | 120 |
Set Membership: |
|