Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01434079_m1
          See other ACTR2 GE Assays ›
          Gene Symbol:
          Actr2
          Gene Name:
          ARP2 actin-related protein 2 homolog (yeast)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.14: 104340783 - 104375648 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01434079_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Actr2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001009268.1 NP_001009268.1 3 - 4 437 83
          GenBank mRNA BC088442.1 - 3 - 4 437 7126325 83
          FQ212547.1 - 3 - 4 436 83
          FQ212814.1 - 4 - 5 401 83
          FQ219838.1 - 3 - 4 403 83
          FQ226083.1 - 3 - 4 383 83
          FQ226422.1 - 3 - 4 386 83
          FQ227488.1 - 3 - 4 435 83
          FQ227830.1 - 3 - 4 386 83
          FQ230874.1 - 3 - 4 421 83
          FQ231828.1 - 3 - 4 388 83
          FQ231874.1 - 3 - 4 421 83
          FQ234230.1 - 3 - 4 388 83
          FQ235071.1 - 3 - 4 408 83
          FQ235201.1 - 3 - 4 403 83

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1400002313.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Actr2

          Panther Classification:

          Molecular Function -

          actin and actin related protein actin or actin-binding cytoskeletal protein cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          establishment or maintenance of cell polarity
          asymmetric cell division
          meiotic chromosome movement towards spindle pole
          cytosolic transport
          actin cytoskeleton organization
          meiotic cytokinesis
          Arp2/3 complex-mediated actin nucleation
          cilium assembly
          meiotic cell cycle
          spindle localization
          actin binding
          structural constituent of cytoskeleton
          ATP binding
          actin filament binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1059422 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 125981 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 351919 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 415436 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 439928 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 19013 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 434555 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 86136 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 485821 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 421930 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 644380 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 19686 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 513988 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 794068 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 510225 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 929560 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 161193 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 350024 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 423777 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 625449 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 645547 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 585346 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 759898 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 582490 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 109794 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 386392 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 271411 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 172940 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 132350 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 46915 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 103351 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 34805 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1059422 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1059422 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1059422 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1059422 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 55880 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 57195 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 17188 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10778612 - -
          Affymetrix Rat Gene 1.0 ST Array 10778608 - -
          Affymetrix Rat Gene 1.0 ST Array 10778609 - -
          Affymetrix Rat Gene 1.0 ST Array 10778610 - -
          Affymetrix Rat Gene 1.0 ST Array 10778606 - -
          Affymetrix Rat Gene 1.0 ST Array 10778607 - -
          Affymetrix Rat Gene 1.0 ST Array 10778611 - -
          Affymetrix Rat Gene 1.1 ST Array 10778605 - -
          Rat Genome 230 2.0 Array 1375424_PM_at - -
          Rat Genome 230 2.0 Array 1384333_PM_at - -
          Rat Genome 230 2.0 Array 1379687_PM_at - -
          Rat Genome 230 2.0 Array 1389904_at - -
          Rat Genome 230 2.0 Array 1389904_PM_at - -
          Rat Genome 230 2.0 Array 1384333_at - -
          Rat Genome 230 2.0 Array 1398626_s_at - -
          Rat Genome 230 2.0 Array 1379687_at - -
          Rat Genome 230 2.0 Array 1375424_at - -
          Rat Genome 230 2.0 Array 1398626_PM_s_at - -
          Affymetrix HT Rat Focus Array 1384333_PM_at - -
          Affymetrix HT Rat Focus Array 1384333_at - -
          Affymetrix HT Rat Focus Array 1398626_s_at - -
          Affymetrix HT Rat Focus Array 1375424_PM_at - -
          Affymetrix HT Rat Focus Array 1398626_PM_s_at - -
          Affymetrix HT Rat Focus Array 1375424_at - -
          Affymetrix Rat Clariom S Assay PSR1400017154.rn.1 TC1400002313.rn.1 RTA TC
          Rat Expression Set 230 1375424_at - -
          Rat Expression Set 230 1375424_PM_at - -
          Rat Expression Set 230 1384333_at - -
          Rat Expression Set 230 1398626_PM_s_at - -
          Rat Expression Set 230 1384333_PM_at - -
          Rat Expression Set 230 1398626_s_at - -
          Affymetrix Rat Gene 1.0 ST Array 10778605 - -
          Rat Genome U34 Set AA801058_at - -
          Rat Genome U34 Set rc_AI237065_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1367256 - -
          RatRef-12 Expression BeadChip ILMN_57628 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline