Search
Search
| Gene Symbol: Aftph | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006251532.2 | XP_006251594.1 | 9 - 10 | 2867 | 71 | |
| XM_006251534.2 | XP_006251596.1 | 9 - 10 | 2704 | 71 | ||
| XM_008770443.1 | XP_008768665.1 | 10 - 11 | 3021 | 71 | ||
| XM_223651.8 | XP_223651.5 | 8 - 9 | 2785 | 71 | ||
| GenBank mRNA | FQ230354.1 | - | 7 - 8 | 976 | 71 | |
Set Membership: |
|