Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01434170_m1
          See other UGP2 GE Assays ›
          Gene Symbol:
          Ugp2
          Gene Name:
          UDP-glucose pyrophosphorylase 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.14: 106208174 - 106249322 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01434170_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ugp2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001024743.1 NP_001019914.1 8 - 9 1563 121
          XM_006251538.1 XP_006251600.1 8 - 9 1515 121
          GenBank mRNA BC084711.1 - 3 - 4 670 121
          BC097369.1 - 8 - 9 1563 7443949 121

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1400002338.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ugp2

          Gene Ontology Categories:

          Function(s) Process(es)

          UDP-glucose metabolic process
          glucose 1-phosphate metabolic process
          UTP:glucose-1-phosphate uridylyltransferase activity
          glucose binding
          pyrimidine ribonucleotide binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P410005 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 853232 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 927548 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 686810 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 409317 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 554955 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 814847 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 171480 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 360947 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 238233 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 219418 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 522047 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 480658 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 142301 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 190767 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 398419 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 478497 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 918985 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 312391 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 515985 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 739424 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 731558 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 30613 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 330494 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 88584 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 678811 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 179935 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 693436 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 211523 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 160258 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 46136 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 75194 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 13510 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 64247 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 63330 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P410005 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P410005 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P410005 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P410005 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P992780 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P136342 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 81367 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 52458 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 87181 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10778648 - -
          Affymetrix Rat Gene 1.0 ST Array 10778657 - -
          Affymetrix Rat Gene 1.0 ST Array 10778651 - -
          Affymetrix Rat Gene 1.0 ST Array 10778653 - -
          Affymetrix Rat Gene 1.0 ST Array 10778649 - -
          Affymetrix Rat Gene 1.0 ST Array 10778650 - -
          Affymetrix Rat Gene 1.0 ST Array 10778655 - -
          Affymetrix Rat Gene 1.0 ST Array 10778654 - -
          Affymetrix Rat Gene 1.0 ST Array 10778658 - -
          Affymetrix Rat Gene 1.0 ST Array 10778656 - -
          Affymetrix Rat Gene 1.0 ST Array 10778652 - -
          Affymetrix Rat Gene 1.1 ST Array 10778647 - -
          Rat Genome 230 2.0 Array 1388410_PM_at - -
          Rat Genome 230 2.0 Array 1388410_at - -
          Affymetrix HT Rat Focus Array 1388410_PM_at - -
          Affymetrix HT Rat Focus Array 1388410_at - -
          Affymetrix Rat Clariom S Assay PSR1400017314.rn.1 TC1400002338.rn.1 RTA TC
          Rat Expression Set 230 1388410_PM_at - -
          Rat Expression Set 230 1388410_at - -
          Affymetrix Rat Gene 1.0 ST Array 10778647 - -
          Rat Genome U34 Set rc_AI059618_at - -
          Rat Genome U34 Set rc_AI170679_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358829 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-t4kr2:80/100.66.77.141:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0