Search
Search
| Gene Symbol: Xpo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053490.1 | NP_445942.1 | 11 - 12 | 1248 | 67 | |
| XM_006251578.2 | XP_006251640.1 | 12 - 13 | 2044 | 67 | ||
| XM_008770437.1 | XP_008768659.1 | 10 - 11 | 1572 | 67 | ||
| GenBank mRNA | AB105193.1 | - | 11 - 12 | 1248 | 67 | |
Set Membership: |
|