Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Rn01434526_g1
See other ETAA1 GE Assays ›
Gene Symbol:
Etaa1
Gene Name:
Ewing tumor-associated antigen 1
Gene Aliases:
RGD1560957
Chromosome Location:
Chr.14: 97196079 - 97199067 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID
Rn01434526_g1
Best Coverage
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
435.00
Your Price
Online offer:
408.65
408.65
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: Etaa1
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001109094.1
NP_001102564.1
424
97
Back To Top
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact
Tech Support
if you need more information.
Set Membership:
Amplicon length between 86 and 100
Detect Genomic DNA
Recommended Assay
Rattus norvegicus
Probe spans exons
MicroArray Probes:
Agilent
Affymetrix
Array Name
Probe IDs
Targets
Target Type
SurePrint G3 Rat Gene Expression Microarray G4131A
A_43_P17761
-
-
Array Name
Probe IDs
Targets
Target Type
Affymetrix Rat Clariom S Assay
PSR1400017026.rn.1
TC1400002290.rn.1
RTA TC
Affymetrix Rat Clariom S Assay
PSR1400017027.rn.1
TC1400002290.rn.1
RTA TC
Affymetrix Rat Clariom S Assay
PSR1400017028.rn.1
TC1400002290.rn.1
RTA TC
Affymetrix Rat Clariom S Assay
PSR1400017025.rn.1
TC1400002290.rn.1
RTA TC
Affymetrix Rat Clariom S Assay
PSR1400017029.rn.1
TC1400002290.rn.1
RTA TC
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit