Search
Search
| Gene Symbol: Eef1e1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106106.1 | NP_001099576.1 | 2 - 3 | 304 | 138 | |
| GenBank mRNA | BC166841.1 | - | 292 | 9027959 | 138 | |
| FQ212823.1 | - | 2 - 3 | 291 | 138 | ||
| FQ219292.1 | - | 2 - 3 | 323 | 138 | ||
| FQ220783.1 | - | 2 - 3 | 305 | 138 | ||
| FQ223579.1 | - | 2 - 3 | 291 | 138 | ||
| FQ225943.1 | - | 2 - 3 | 323 | 138 | ||
| FQ229476.1 | - | 2 - 3 | 138 | |||
| FQ231336.1 | - | 2 - 3 | 291 | 138 | ||
| FQ233549.1 | - | 2 - 3 | 291 | 138 | ||
| FQ234239.1 | - | 2 - 3 | 303 | 138 | ||
Set Membership: |
|