Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01434827_m1
          See other PRPF4B GE Assays ›
          Gene Symbol:
          Prpf4b
          Gene Name:
          pre-mRNA processing factor 4B
          Gene Aliases:
          -
          Chromosome Location:
          Chr.17: 30680454 - 30714542 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01434827_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Prpf4b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001011923.1 NP_001011923.1 11 - 12 2511 118
          GenBank mRNA BC085927.1 - 11 - 12 2511 7110150 118

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1700001637.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Prpf4b

          Gene Ontology Categories:

          Function(s) Process(es)

          mRNA processing
          protein phosphorylation
          RNA splicing
          protein serine/threonine kinase activity
          ATP binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1016170 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 719033 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 20619 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 973927 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 60031 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 122670 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 661488 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 375886 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 335080 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 82849 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 629866 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 513328 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 939234 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 825319 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 893188 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 932006 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 157701 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 272034 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 667111 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 499793 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 440937 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 210827 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 175567 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 50763 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 712126 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 871770 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 92315 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 81378 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 41767 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1016170 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1016170 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1016170 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1033684 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1016170 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 71298 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 34233 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 11952 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 17486 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 21457 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 77642 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10798082 - -
          Affymetrix Rat Gene 1.0 ST Array 10798092 - -
          Affymetrix Rat Gene 1.0 ST Array 10798081 - -
          Affymetrix Rat Gene 1.0 ST Array 10798078 - -
          Affymetrix Rat Gene 1.0 ST Array 10798088 - -
          Affymetrix Rat Gene 1.0 ST Array 10798086 - -
          Affymetrix Rat Gene 1.0 ST Array 10798087 - -
          Affymetrix Rat Gene 1.0 ST Array 10798084 - -
          Affymetrix Rat Gene 1.0 ST Array 10798089 - -
          Affymetrix Rat Gene 1.0 ST Array 10798090 - -
          Affymetrix Rat Gene 1.0 ST Array 10798077 - -
          Affymetrix Rat Gene 1.0 ST Array 10798083 - -
          Affymetrix Rat Gene 1.0 ST Array 10798085 - -
          Affymetrix Rat Gene 1.0 ST Array 10798076 - -
          Affymetrix Rat Gene 1.0 ST Array 10798091 - -
          Affymetrix Rat Gene 1.0 ST Array 10798080 - -
          Affymetrix Rat Gene 1.1 ST Array 10798074 - -
          Rat Genome 230 2.0 Array 1382646_at - -
          Rat Genome 230 2.0 Array 1397203_PM_at - -
          Rat Genome 230 2.0 Array 1376010_PM_at - -
          Rat Genome 230 2.0 Array 1382646_PM_at - -
          Rat Genome 230 2.0 Array 1376010_at - -
          Rat Genome 230 2.0 Array 1397203_at - -
          Affymetrix HT Rat Focus Array 1376010_at - -
          Affymetrix HT Rat Focus Array 1397203_PM_at - -
          Affymetrix HT Rat Focus Array 1397203_at - -
          Affymetrix HT Rat Focus Array 1382646_PM_at - -
          Affymetrix HT Rat Focus Array 1382646_at - -
          Affymetrix HT Rat Focus Array 1376010_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1700011354.rn.1 TC1700001637.rn.1 RTA TC
          Rat Expression Set 230 1376010_PM_at - -
          Rat Expression Set 230 1376010_at - -
          Rat Expression Set 230 1382646_at - -
          Rat Expression Set 230 1382646_PM_at - -
          Rat Expression Set 230 1397203_at - -
          Rat Expression Set 230 1397203_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10798074 - -
          Rat Genome U34 Set rc_AA955630_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22293200 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376006 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376742 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline