Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01434847_m1
          See other PRL8A2 GE Assays ›
          Gene Symbol:
          Prl8a2
          Gene Name:
          prolactin family 8, subfamily a, member 2
          Gene Aliases:
          Dprp, Dtprp, d/tPRP
          Chromosome Location:
          Chr.17: 39563383 - 39575777 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01434847_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Prl8a2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022846.2 NP_074037.1 5 - 6 535 92
          XM_006253906.1 XP_006253968.1 5 - 6 595 92
          GenBank mRNA BC097316.1 - 5 - 6 536 7442457 92
          FQ219955.1 - 5 - 6 595 92
          FQ220110.1 - 5 - 6 595 92
          FQ222092.1 - 1 - 2 81 92
          FQ228468.1 - 1 - 2 165 92
          FQ229664.1 - 5 - 6 569 92
          FQ229702.1 - 5 - 6 593 92
          FQ229882.1 - 5 - 6 595 92
          L06441.1 - 5 - 6 537 92

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1700001749.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Prl8a2

          Panther Classification:

          Molecular Function -

          peptide hormone

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          female pregnancy
          maternal process involved in female pregnancy
          hormone activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P192950 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 844986 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 727871 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 40728 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 950516 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 331343 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 295846 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 483564 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 123259 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 126399 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P192950 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P192950 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P192950 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P192950 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 99432 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10798268 - -
          Affymetrix Rat Gene 1.0 ST Array 10798270 - -
          Affymetrix Rat Gene 1.0 ST Array 10798266 - -
          Affymetrix Rat Gene 1.0 ST Array 10798269 - -
          Affymetrix Rat Gene 1.0 ST Array 10798265 - -
          Affymetrix Rat Gene 1.0 ST Array 10798267 - -
          Affymetrix Rat Gene 1.1 ST Array 10798264 - -
          Rat Genome 230 2.0 Array 1368715_s_at - -
          Rat Genome 230 2.0 Array 1368714_PM_at - -
          Rat Genome 230 2.0 Array 1368714_at - -
          Rat Genome 230 2.0 Array 1368715_PM_s_at - -
          Affymetrix HT Rat Focus Array 1368714_PM_at - -
          Affymetrix HT Rat Focus Array 1368715_s_at - -
          Affymetrix HT Rat Focus Array 1368714_at - -
          Affymetrix HT Rat Focus Array 1368715_PM_s_at - -
          Affymetrix Rat Clariom S Assay PSR1700011889.rn.1 TC1700001749.rn.1 RTA TC
          Rat Expression Set 230 1368715_PM_s_at - -
          Rat Expression Set 230 1368714_PM_at - -
          Rat Expression Set 230 1368714_at - -
          Rat Expression Set 230 1368715_s_at - -
          Affymetrix Rat Gene 1.0 ST Array 10798264 - -
          Rat Genome U34 Set L06441_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20876239 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1351507 - -
          RatRef-12 Expression BeadChip ILMN_62548 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline