Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01434874_s1
          See other NFIL3 GE Assays ›
          Gene Symbol:
          Nfil3
          Gene Name:
          nuclear factor, interleukin 3 regulated
          Gene Aliases:
          E4BP4
          Chromosome Location:
          Chr.17: 12261102 - 12276316 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01434874_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nfil3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053727.2 NP_446179.2 2 1923 83
          XM_006253667.2 XP_006253729.1 2 2597 83
          XM_006253668.2 XP_006253730.1 2 1978 83
          XM_006253669.2 XP_006253731.1 2 3160 83
          XM_006253670.2 XP_006253732.1 2 2332 83
          GenBank mRNA AY004663.1 - 1 1759 83
          BC072527.1 - 2 1923 7097417 83

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nfil3

          Panther Classification:

          Molecular Function -

          basic leucine zipper transcription factor DNA-binding transcription factor gene-specific transcriptional regulator

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          immune response
          circadian rhythm
          positive regulation of gene expression
          cellular response to interleukin-4
          RNA polymerase II regulatory region sequence-specific DNA binding
          RNA polymerase II core promoter sequence-specific DNA binding
          transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
          transcription factor activity, sequence-specific DNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P201028 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 184865 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 764905 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 105673 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 389564 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 156259 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843389 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 338707 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 202400 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 343498 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 899019 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 107319 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 114532 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P201028 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P201028 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P201028 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P201028 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 54988 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 60575 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10794228 - -
          Affymetrix Rat Gene 1.0 ST Array 10794226 - -
          Affymetrix Rat Gene 1.0 ST Array 10794227 - -
          Affymetrix Rat Gene 1.1 ST Array 10794225 - -
          Rat Genome 230 2.0 Array 1368488_PM_at - -
          Rat Genome 230 2.0 Array 1368488_at - -
          Affymetrix HT Rat Focus Array 1368488_at - -
          Affymetrix HT Rat Focus Array 1368488_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1700001745.rn.1 TC1700000212.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700001740.rn.1 TC1700000212.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700001746.rn.1 TC1700000212.rn.1 RTA TC
          Rat Expression Set 230 1368488_at - -
          Rat Expression Set 230 1368488_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10794225 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21240715 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1360188 - -
          RatRef-12 Expression BeadChip ILMN_68178 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline